missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,6-diaminohexane, 98%

Catalog No. AAL1994814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1994814 25 g
AAL1994803 1 g
AAL1994806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1994814 Supplier Thermo Scientific Chemicals Supplier No. L1994814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51857-17-1 | C11H24N2O2 | 216.325 g/mol

N-Boc-1,6-diaminohexane is used to prepare 1,3-Bis[6-(Boc-amino)hexyl]urea by reacting with carbonyl dichloride in the presence of triethylamine. Further, it is used as a reagent for the introduction of a C6-spacer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-1,6-diaminohexane is used to prepare 1,3-Bis[6-(Boc-amino)hexyl]urea by reacting with carbonyl dichloride in the presence of triethylamine. Further, it is used as a reagent for the introduction of a C6-spacer.

Solubility
Soluble in dichloromethane and ethyl acetate.

Notes
Air sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 51857-17-1
Molecular Formula C11H24N2O2
Molecular Weight (g/mol) 216.325
MDL Number MFCD00671489
InChI Key RVZPDKXEHIRFPM-UHFFFAOYSA-N
Synonym n-boc-1,6-diaminohexane, n-boc-1,6-hexanediamine, tert-butyl n-6-aminohexyl carbamate, tert-butyl 6-aminohexyl carbamate, n-tert-butoxycarbonyl-1,6-diaminohexane, n-tert-butoxycarbonyl-1,6-hexanediamine, tert-butyl 6-aminohexylcarbamate, boc-1,6-diaminohexane hydrochloride, 6-tert-butoxycarbonylamino hexylamine, n-6-aminohexyl carbamic acid tert-butyl ester
PubChem CID 2733170
IUPAC Name tert-butyl N-(6-aminohexyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCCCN

Specifications

Boiling Point 106°C to 110°C (0.3mmHg)
Refractive Index 1.461
UN Number UN2735
Beilstein 2089264
Solubility Information Soluble in dichloromethane and ethyl acetate.
Density 0.965
Quantity 25 g
Flash Point >110°C (230°F)
Formula Weight 216.33
Sensitivity Air Sensitive
Percent Purity 98%
Chemical Name or Material N-Boc-1,6-diaminohexane
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 628-984-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only