missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-glutamic acid, 99%

Catalog No. AAA1768618
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1768618 50 g
AAA1768609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1768618 Supplier Thermo Scientific Chemicals Supplier No. A1768618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1155-62-0 | C13H15NO6 | 281.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1155-62-0
Molecular Formula C13H15NO6
Molecular Weight (g/mol) 281.264
MDL Number MFCD00002801
InChI Key PVFCXMDXBIEMQG-JTQLQIEISA-N
Synonym n-cbz-l-glutamic acid, z-glu-oh, n-benzyloxycarbonyl-l-glutamic acid, carbobenzoxy-l-glutamic acid, n-carbobenzoxy-l-glutamic acid, s-2-benzyloxy carbonyl amino pentanedioic acid, n-benzyloxycarbonylglutamic acid, n-carbobenzyloxy-l-glutamic acid, z-l-glu-oh
PubChem CID 70866
IUPAC Name (2S)-2-(phenylmethoxycarbonylamino)pentanedioic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CCC(=O)O)C(=O)O

Specifications

Melting Point 108°C to 116°C
Quantity 50 g
Beilstein 2061272
Optical Rotation −7.5° (c=4 in Acetic Acid)
Formula Weight 281.27
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid
EINECSNumber 214-584-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only