missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylglycine ethyl ester, 97%

CAS: 6436-90-4 | C11H15NO2 | 193.25 g/mol

$123.04 - $1071.53

Chemical Identifiers

CAS 6436-90-4
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.25
MDL Number MFCD00009174
InChI Key ULOLIZHBYWAICY-UHFFFAOYSA-N
Synonym n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester
PubChem CID 80908
IUPAC Name ethyl 2-(benzylamino)acetate
SMILES CCOC(=O)CNCC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1549614
View Documents
Thermo Scientific Chemicals
L1549614
25 g
Each for $123.04
Only null left
 
AAL1549622
View Documents
Thermo Scientific Chemicals
L1549622
100 g
Each for $353.05
Only null left
 
AAL1549636
View Documents
Thermo Scientific Chemicals
L1549636
500 g
Each for $1,071.53
Only null left
 
Description

Description

N-Benzylglycine ethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzylglycine ethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

6436-90-4
193.25
ULOLIZHBYWAICY-UHFFFAOYSA-N
80908
CCOC(=O)CNCC1=CC=CC=C1
C11H15NO2
MFCD00009174
n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester
ethyl 2-(benzylamino)acetate

Specifications

6436-90-4
C11H15NO2
MFCD00009174
n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester
ULOLIZHBYWAICY-UHFFFAOYSA-N
CCOC(=O)CNCC1=CC=CC=C1
193.25
112°C (233°F)
193.25
N-Benzylglycine ethyl ester
102°C to 104°C (1mmHg)
1.5053
2104816
Not miscible or difficult to mix in water.
1.048
ethyl 2-(benzylamino)acetate
25 g
80908
97%
Videos
Safety and Handling

Safety and Handling

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 229-218-6

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only