missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylglycine ethyl ester, 97%

CAS: 6436-90-4 | C11H15NO2 | 193.25 g/mol

Supplier:  Thermo Scientific Chemicals L1549622

 View more versions of this product

Catalog No. AAL1549622


Only null left
Explore available promotions

Description

Description

N-Benzylglycine ethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzylglycine ethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Specifications

N-Benzylglycine ethyl ester
102°C to 104°C (1mmHg)
1.5053
2104816
Not miscible or difficult to mix in water.
1.048
ethyl 2-(benzylamino)acetate
100 g
80908
97%
6436-90-4
C11H15NO2
MFCD00009174
n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester
ULOLIZHBYWAICY-UHFFFAOYSA-N
CCOC(=O)CNCC1=CC=CC=C1
193.25
112°C (233°F)
193.25

Chemical Identifiers

6436-90-4
193.25
ULOLIZHBYWAICY-UHFFFAOYSA-N
80908
CCOC(=O)CNCC1=CC=CC=C1
C11H15NO2
MFCD00009174
n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester
ethyl 2-(benzylamino)acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 229-218-6

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only