accessibility menu, dialog, popup

UserName

H3COONH

CAS RN 6436-90-4

IUPAC Name: ethyl 2-(benzylamino)acetate
Synonyms: ethyl 2-(benzylamino)acetate
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: ULOLIZHBYWAICY-UHFFFAOYSA-N
SMILES: CCOC(=O)CNCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)