Learn More
N-Benzylglycine ethyl ester, 97%
CAS: 6436-90-4 | C11H15NO2 | 193.25 g/mol
Supplier: Thermo Scientific Chemicals L1549636
Description
N-Benzylglycine ethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Benzylglycine ethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
| N-Benzylglycine ethyl ester | |
| 102°C to 104°C (1mmHg) | |
| 1.5053 | |
| 2104816 | |
| Not miscible or difficult to mix in water. | |
| 1.048 | |
| ethyl 2-(benzylamino)acetate | |
| 500 g | |
| 80908 | |
| 97% |
| 6436-90-4 | |
| C11H15NO2 | |
| MFCD00009174 | |
| n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester | |
| ULOLIZHBYWAICY-UHFFFAOYSA-N | |
| CCOC(=O)CNCC1=CC=CC=C1 | |
| 193.25 | |
| 112°C (233°F) | |
| 193.25 |
Chemical Identifiers
| 6436-90-4 | |
| 193.25 | |
| ULOLIZHBYWAICY-UHFFFAOYSA-N | |
| 80908 | |
| CCOC(=O)CNCC1=CC=CC=C1 |
| C11H15NO2 | |
| MFCD00009174 | |
| n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester | |
| ethyl 2-(benzylamino)acetate |
Safety and Handling
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 229-218-6
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only