missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylbenzamide, 99%

Catalog No. AC191170250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC191170250 25 g Plastic bottle
AC191170050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC191170250 Supplier Thermo Scientific Chemicals Supplier No. 191170250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1485-70-7 | C14H13NO | 211.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1485-70-7
Molecular Formula C14H13NO
Molecular Weight (g/mol) 211.26
MDL Number MFCD00003070
InChI Key LKQUCICFTHBFAL-UHFFFAOYSA-N
Synonym benzamide, n-phenylmethyl, n-benzyl-benzamide, phenyl-n-benzylcarboxamide, benzoylbenzylamine, n∼1∼-benzylbenzamide, benzamide, n-benzyl, n-phenylmethyl benzamide, acmc-1bx9y, n-benzylbenzamide, lkqucicfthbfal-uhfffaoysa
PubChem CID 73878
IUPAC Name N-benzylbenzamide
SMILES O=C(NCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 104.0°C to 107.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H5CONHCH2C6H5
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 211.26
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material N-Benzylbenzamide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face prot
EINECSNumber 216-063-4

RUO – Research Use Only