missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzoyl-D-glucosamine 98.0+%, TCI America™
Spécifications
Spécifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 283.28 |
| Percent Purity | ≥98.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Benzoyl-D-glucosamine |
Sécurité et manipulation
| TSCA | No |