missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N(alpha)-Boc-L-asparagine, 98+%

Numéro de catalogue. AAA1601906
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1601906 5 g
AAA1601914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAA1601906 Fournisseur Thermo Scientific Chemicals Code fournisseur A1601906
Il en reste null

CAS: 7536-55-2 | C9H16N2O5 | 232.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 7536-55-2
Molecular Formula C9H16N2O5
Molecular Weight (g/mol) 232.236
MDL Number MFCD00038152
InChI Key FYYSQDHBALBGHX-YFKPBYRVSA-N
Synonym boc-asn-oh, boc-l-asparagine, boc-asn, n-tert-butoxycarbonyl-l-asparagine, tert-butoxycarbonylasparagine, nalpha-tert-butoxycarbonyl-l-asparagine, boc-asparagine, tert-butoxycarbonyl-l-asparagine, n-tert-butoxycarbonyl asparagine, tert-butyloxycarbonyl-l-asparagine
PubChem CID 82035
ChEBI CHEBI:3146
IUPAC Name (2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O

Spécifications

Melting Point ∼170°C (decomposition)
Beilstein 1977963
Quantity 5 g
Formula Weight 232.24
Optical Rotation −7.3° (c=2 in DMF)
Percent Purity ≥98%
Chemical Name or Material N(alpha)-Boc-L-asparagine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-405-2
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only