missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N(alpha)-Boc-D-tryptophan, 97%

Catalog No. AAL0921406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0921406 5 g
AAL0921403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0921406 Supplier Thermo Scientific Chemicals Supplier No. L0921406

CAS: 5241-64-5 | C16H20N2O4 | 304.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5241-64-5
Molecular Formula C16H20N2O4
Molecular Weight (g/mol) 304.35
MDL Number MFCD00037944
InChI Key NFVNYBJCJGKVQK-CYBMUJFWSA-N
Synonym boc-d-trp-oh, n-boc-d-tryptophan, n-tert-butoxy carbonyl-d-tryptophan, n-tert-butoxycarbonyl-d-tryptophan, boc-d-tryptophan, boc-d-trp, nalpha-boc-d-tryptophan, boc-d-tryptophane, nalpha-boc-d-tryptophane, tert-butoxycarbonyl
PubChem CID 111050
IUPAC Name (2R)-3-(1H-indol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point ∽138°C (decomposition)
Beilstein 4237334
Quantity 5 g
Formula Weight 304.35
Optical Rotation +21° (c=1 in Acetic Acid)
Percent Purity 97%
Chemical Name or Material N(alpha)-Boc-D-tryptophan
EINECSNumber 226-042-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only