missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Benzyloxycarbonyl-L-histidine, 98+%

Catalog No. AAL0911406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0911406 5 g
AAL0911403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0911406 Supplier Thermo Scientific Chemicals Supplier No. L0911406
Only null left

CAS: 14997-58-1 | C14H15N3O4 | 289.291 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14997-58-1
Molecular Formula C14H15N3O4
Molecular Weight (g/mol) 289.291
MDL Number MFCD00065960
InChI Key WCOJOHPAKJFUDF-LBPRGKRZSA-N
Synonym z-his-oh, n-cbz-l-histidine, cbz-his-oh, cbz-l-his-oh, nalpha-cbz-l-histidine, cbz-histidine, nalpha-carbobenzoxy-l-histidine, l-histidine, n-phenylmethoxy carbonyl, z-l-histidine, s-2-benzyloxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid
PubChem CID 736313
IUPAC Name (2S)-3-(1H-imidazol-5-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC2=CN=CN2)C(=O)O

Specifications

Melting Point ∽165°C (decomposition)
Beilstein 3561682
Quantity 5 g
Formula Weight 289.29
Optical Rotation −24° (c=6 in 6N HCl)
Percent Purity ≥98%
Chemical Name or Material N(alpha)-Benzyloxycarbonyl-L-histidine
EINECSNumber 239-084-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only