missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Benzyloxycarbonyl-L-glutamine, 99%

Catalog No. AAA1824022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1824022 100 g
AAA1824006 5 g
AAA1824014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1824022 Supplier Thermo Scientific Chemicals Supplier No. A1824022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2650-64-8 | C13H16N2O5 | 280.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2650-64-8
Molecular Formula C13H16N2O5
Molecular Weight (g/mol) 280.28
MDL Number MFCD00008043
InChI Key JIMLDJNLXLMGLX-JTQLQIEISA-N
Synonym z-gln-oh, n-carbobenzyloxy-l-glutamine, cbz-l-glutamine, cbz-gln-oh, n-carbobenzoxy-l-glutamine, benzyloxycarbonyl-l-glutamine, carbobenzoxyglutamine, n-cbz-l-glutamine, cbz-l-gln-oh, carbobenzoxy-l-glutamine
PubChem CID 75855
IUPAC Name (2S)-5-amino-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES NC(=O)CC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 134°C to 137°C
Beilstein 2061271
Quantity 100 g
Formula Weight 280.28
Optical Rotation −7° (c=2 in Ethanol)
Percent Purity 99%
Chemical Name or Material N(alpha)-Benzyloxycarbonyl-L-glutamine
EINECSNumber 220-173-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only