missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-alpha-Acetyl-L-glutamine, 99%

Catalog No. AAL0678030
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0678030 250 g
AAL0678018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0678030 Supplier Thermo Scientific Chemicals Supplier No. L0678030

CAS: 2490-97-3 | C7H12N2O4 | 188.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2490-97-3
Molecular Formula C7H12N2O4
Molecular Weight (g/mol) 188.183
MDL Number MFCD00038159
InChI Key KSMRODHGGIIXDV-YFKPBYRVSA-N
Synonym n-acetyl-l-glutamine, aceglutamide, l-glutamine, n-acetyl, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid, acetylglutamine, n-alpha-acetyl-l-glutamine, n-acetylglutamine, acutil-s, acetyl-l-glutamine
PubChem CID 182230
ChEBI CHEBI:21553
IUPAC Name (2S)-2-acetamido-5-amino-5-oxopentanoic acid
SMILES CC(=O)NC(CCC(=O)N)C(=O)O

Specifications

Melting Point ∽206°C (decomposition)
Beilstein 1727471
Merck Index 14,25
Quantity 250 g
Formula Weight 188.18
Optical Rotation −12° (c=3 in Water)
Percent Purity 99%
Chemical Name or Material N-alpha-Acetyl-L-glutamine
EINECSNumber 219-647-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only