Learn More
N-Allylaniline, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 102871000
| Quantity | 100g |
|---|
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C9H11N | |
| MFCD00008638 | |
| n-allylaniline, n-allyl-aniline, benzenamine, n-2-propenyl, unii-q7s639gwxg, n-prop-2-en-1-ylaniline, q7s639gwxg, phenylprop-2-enylamine, nyl, allylphenylamine, allyl-phenyl-amine | |
| N-prop-2-enylaniline |
Specifications
| N-Allylaniline | |
| 589-09-3 | |
| 100.0 | |
| 218°C to 220°C | |
| Authentic | |
| Glass bottle | |
| 1.5620 to 1.564 | |
| 100g | |
| 12, 170 | |
| n-allylaniline, n-allyl-aniline, benzenamine, n-2-propenyl, unii-q7s639gwxg, n-prop-2-en-1-ylaniline, q7s639gwxg, phenylprop-2-enylamine, nyl, allylphenylamine, allyl-phenyl-amine | |
| C=CCNC1=CC=CC=C1 | |
| 133.19 | |
| 133.19 |
| 95% | |
| 94.0 | |
| 0.9820g/mL | |
| 89°C | |
| 95% | |
| C9H11N | |
| C6H5NHCH2CH=CH2 | |
| MFCD00008638 | |
| 0.982 | |
| LQFLWKPCQITJIH-UHFFFAOYSA-N | |
| N-prop-2-enylaniline | |
| 68525 | |
| 95% |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 209-633-9
Promotions