missing translation for 'onlineSavingsMsg'
Learn More

N-Allyl-N'-(2-hydroxyethyl)thiourea, 97%

Catalog No. AAA1816130
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1816130 250 g
AAA1816118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1816130 Supplier Thermo Scientific Chemicals Supplier No. A1816130
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 105-81-7 | C6H12N2OS | 160.235 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105-81-7
Molecular Formula C6H12N2OS
Molecular Weight (g/mol) 160.235
MDL Number MFCD00002838
InChI Key VUVPNTYTOUGMDG-UHFFFAOYSA-N
Synonym 1-allyl-3-2-hydroxyethyl-2-thiourea, 1-allyl-3-2-hydroxyethyl thiourea, thiourea, n-2-hydroxyethyl-n'-2-propenyl, n-allyl-n'-2-hydroxyethyl thiourea, n-2-hydroxyethyl-n'-allylthiourea, n-2-hydroxyethyl-n'-2-propenylthiourea, n-allyl-n'-2-hydroxyethylthiourea, urea, 1-allyl-3-2-hydroxyethyl-2-thio, n-allyl-n'-beta-hydroxyethyl thiourea
PubChem CID 2735273
IUPAC Name 1-(2-hydroxyethyl)-3-prop-2-enylthiourea
SMILES C=CCNC(=S)NCCO

Specifications

Melting Point 77°C to 79°C
Quantity 250 g
Formula Weight 160.24
Percent Purity 97%
Chemical Name or Material N-Allyl-N'-(2-hydroxyethyl)thiourea
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-334-7
RTECSNumber YR7913000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only