missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylethylenediamine, tech. 90%

Catalog No. AAL1339806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1339806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1339806 Supplier Thermo Scientific Chemicals Supplier No. L1339806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1001-53-2 | C4H10N2O | 102.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1001-53-2
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.137
MDL Number MFCD00008163
InChI Key DAKZISABEDGGSV-UHFFFAOYSA-N
Synonym n-2-aminoethyl acetamide, n-acetylethylenediamine, acetamide, n-2-aminoethyl, n-2-amino-ethyl-acetamide, unii-fql33v3kzd, fql33v3kzd, acetylethylenediamine, 2-acetamidoethylamine, acetylethylene diamine, n-acetylethylendiamine
PubChem CID 66082
IUPAC Name N-(2-aminoethyl)acetamide
SMILES CC(=O)NCCN

Specifications

Melting Point 44°C to 50°C
Boiling Point 128°C (3 mmHg)
Flash Point >110°C (230°F)
Linear Formula CH3CONHCH2CH2NH2
Quantity 5 g
UN Number UN3259
Beilstein 1743120
Sensitivity Hygroscopic
Formula Weight 102.14
Percent Purity 90%
Grade Technical
Chemical Name or Material N-Acetylethylenediamine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only