missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-S-trityl-L-cysteine, 95%

Catalog No. AAH6668906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6668906 5 g
AAH6668903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6668906 Supplier Thermo Scientific Chemicals Supplier No. H6668906
Only null left

CAS: 27486-87-9 | C24H23NO3S | 405.512 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27486-87-9
Molecular Formula C24H23NO3S
Molecular Weight (g/mol) 405.512
MDL Number MFCD00236747
InChI Key KCVPASSMLHHOIF-QFIPXVFZSA-N
Synonym ac-cys trt-oh, n-acetyl-s-trityl-l-cysteine, r-n-acetyl-s-tritylcysteine, 2r-2-acetamido-3-triphenylmethyl sulfanyl propanoic acid, ac-cys trt-oh;, n-acetyl-s-trityl-l-cysteine;, l-cysteine,n-acetyl-s-triphenylmethyl, l-cysteine, n-acetyl-s-triphenylmethyl, s-trityl-l-cysteine stlc analogue, 37, 2r-2-acetamido-3-tritylsulfanylpropanoic acid
PubChem CID 7408363
IUPAC Name (2R)-2-acetamido-3-tritylsulfanylpropanoic acid
SMILES CC(=O)NC(CSC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O

Specifications

Beilstein 6877409
Quantity 5 g
Formula Weight 405.51
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Acetyl-S-trityl-L-cysteine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only