missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-ornithine, 95%

Catalog No. AAH66648MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66648MD 250 mg
AAH6664803 1 g
AAH6664806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66648MD Supplier Thermo Scientific Chemicals Supplier No. H66648MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6205-08-9 | C7H14N2O3 | 174.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6205-08-9
Molecular Formula C7H14N2O3
Molecular Weight (g/mol) 174.20
MDL Number MFCD00065115
InChI Key JRLGPAXAGHMNOL-UHFFFAOYNA-N
PubChem CID 439232
ChEBI CHEBI:16543
IUPAC Name (2S)-2-acetamido-5-aminopentanoic acid
SMILES CC(=O)NC(CCCN)C(=O)O

Specifications

Synonym n-acetylornithine, ac-orn-oh, n2-acetyl-l-ornithine, n 2-acetyl-l-ornithine, acetylornithine, 2s-2-acetamido-5-aminopentanoic acid, n∼2∼-acetyl-l-ornithine, aor, ornithine, n2-acetyl, nalpha-acetyl-l-ornithine
Quantity 250 mg
Formula Weight 174.2
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Acetyl-L-ornithine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only