missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-methionine, 99%

Catalog No. AAB2431206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2431206 5 g
AAB2431214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2431206 Supplier Thermo Scientific Chemicals Supplier No. B2431206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 65-82-7 | C7H13NO3S | 191.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65-82-7
Molecular Formula C7H13NO3S
Molecular Weight (g/mol) 191.25
MDL Number MFCD00064441
InChI Key XUYPXLNMDZIRQH-LURJTMIESA-N
Synonym n-acetyl-l-methionine, n-acetylmethionine, acetyl-l-methionine, ac-met-oh, l-n-acetyl methionine, acetylmethionine, methionamine, methionine, n-acetyl-, l, thiomedon, l-methionine, n-acetyl
PubChem CID 448580
ChEBI CHEBI:21557
IUPAC Name (2S)-2-acetamido-4-methylsulfanylbutanoic acid
SMILES CSCC[C@H](NC(C)=O)C(O)=O

Specifications

Melting Point 103°C to 107°C
Beilstein 1725552
Merck Index 14,96
Quantity 5 g
Formula Weight 191.25
Optical Rotation −21° (c=1.3 in Water)
Percent Purity 99%
Chemical Name or Material N-Acetyl-L-methionine
EINECSNumber 200-617-7
RTECSNumber PD0480000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only