missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-leucine 99.0+%, TCI America™

Catalog No. A00985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A00985G 5 g
A009825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A00985G Supplier TCI America Supplier No. A00985G

Chemical Identifiers

CAS 1188-21-2
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00065131
InChI Key WXNXCEHXYPACJF-JLDDOWRYNA-N
Synonym n-acetyl-l-leucine, acetyl-l-leucine, ac-leu-oh, n-acetylleucine, s-2-acetamido-4-methylpentanoic acid, n-acetyl-leu, 2s-2-acetamido-4-methylpentanoic acid, n-acetyl-leucine, leucine, n-acetyl-, l, n-acetyl-l-leucin
PubChem CID 70912
ChEBI CHEBI:17786
IUPAC Name (2S)-2-acetamido-4-methylpentanoic acid
SMILES CC(C)C[C@H](NC(C)=O)C(O)=O

Specifications

Melting Point 185°C
Color White
Quantity 5 g
Formula Weight 173.21
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-leucine