missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-leucine, 99%

Catalog No. AC347010250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC347010250 25 g Glass bottle
AC347010050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC347010250 Supplier Thermo Scientific Chemicals Supplier No. 347010250

CAS: 1188-21-2 | C8H15NO3 | 173.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1188-21-2
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00065131
InChI Key WXNXCEHXYPACJF-JLDDOWRYNA-N
Synonym n-acetyl-l-leucine, acetyl-l-leucine, ac-leu-oh, n-acetylleucine, s-2-acetamido-4-methylpentanoic acid, n-acetyl-leu, 2s-2-acetamido-4-methylpentanoic acid, n-acetyl-leucine, leucine, n-acetyl-, l, n-acetyl-l-leucin
PubChem CID 70912
ChEBI CHEBI:17786
IUPAC Name (2S)-2-acetamido-4-methylpentanoic acid
SMILES CC(C)C[C@H](NC(C)=O)C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 190.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CH(NHCOCH3)COOH
Quantity 25 g
Specific Rotation − 24.50
Solubility Information (5% in ethanol) Clear colorless
Formula Weight 173.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-leucine, 99%
Show More Show Less
EINECSNumber 214-706-3
TSCA TSCA

RUO – Research Use Only