missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-isoleucine, 98%

Catalog No. AAH6677114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6677114 25 g
AAH6677106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6677114 Supplier Thermo Scientific Chemicals Supplier No. H6677114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3077-46-1 | C8H15NO3 | 173.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3077-46-1
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00066072
InChI Key JDTWZSUNGHMMJM-IFSCUNPJNA-N
Synonym n-acetyl-l-isoleucine, 2s,3s-2-acetamido-3-methylpentanoic acid, ac-ile-oh, l-isoleucine, n-acetyl, unii-p9cl0y0bvz, p9cl0y0bvz, isoleucine, n-acetyl-, l, acetyl isoleucine, n-acetyl-isoleucine, acetyl-l-isoleucine
PubChem CID 7036275
ChEBI CHEBI:21555
SMILES CC[C@H](C)[C@H](NC(C)=O)C(O)=O

Specifications

Color White
IUPAC Name 2-acetamido-3-methylpentanoic acid
Quantity 25 g
Formula Weight 173.21
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material N-Acetyl-L-isoleucine
EINECSNumber 221-364-9
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only