missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-histidine Monohydrate 99.0+%, TCI America™

Catalog No. A087325G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A087325G 25 g
A08731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A087325G Supplier TCI America Supplier No. A087325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39145-52-3
Molecular Formula C8H13N3O4
Molecular Weight (g/mol) 215.209
MDL Number MFCD00149320
InChI Key PSWSDQRXCOJSFC-FJXQXJEOSA-N
Synonym ac-his-oh.h2o, n-acetyl-l-histidine hydrate, n-acetyl-l-histidine monohydrate, unii-pq2tc3x11o, s-2-acetamido-3-1h-imidazol-4-yl propanoic acid hydrate, pq2tc3x11o, acetyl histidine, 2s-2-acetylamino-3-1h-imidazol-4-yl propanoic acid hydrate, l-n-acetylhistidine monohydrate
PubChem CID 2724380
IUPAC Name (2S)-2-acetamido-3-(1H-imidazol-5-yl)propanoic acid;hydrate
SMILES CC(=O)NC(CC1=CN=CN1)C(=O)O.O

Specifications

Melting Point 163°C
Color White
Quantity 25 g
Formula Weight 197.19
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-histidine Monohydrate