missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-cysteine, 98+%

N-Acetyl-L-cysteine (acetylcysteine, CAS # 616-91-1) is a product of the amino acid L-cysteine.

Supplier:  Thermo Scientific Chemicals A1540922

Quantity 100 g

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA1540922


Il en reste null

Description

Description

A mucolytic agent for isolation of mycobacteria from sputum.N-Acetyl-L-cysteine is used in the liquefaction and decontamination of sputum during the preparation for recovery of mycobacterium, which is called as Petroff's method. It shows antiviral activity against the influenza A viruses. It is used in the treatment of interstitial lung disease to prevent disease progression. It is an antioxidant, dietary supplement and protects liver. Further, it forms complexes with palladium, thereby it dissolves in water and remove palladium from drugs. Further, it is used in the treatment of paracetamol overdose. In addition to this, it is involved in the treatment of autism, Alzheimer's disease, bipolar disorder, major depressive disorder, obsessive-compulsive disorder, schizophrenia, and specific drug addictions like cocaine and cannabis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • N-Acetyl-L-cysteine (acetylcysteine) is a precursor of the antioxidant glutathione, or GSH
  • N-Acetyl-L-cysteine is cytoprotective and may prevent cancer, neurodegeneration, and other disease

Application

  • N-Acetyl-L-cysteine is useful for cellular redox study because of its anti-inflammatory properties
  • It may also be useful for psychiatric research and the study of neurodegeneration
  • It is suitable for in vivo and in vitro experiments
Spécifications

Identifiants chimiques

616-91-1
163.191
PWKSKIMOESPYIA-BYPYZUCNSA-N
12035
(2R)-2-acetamido-3-sulfanylpropanoic acid
C5H9NO3S
MFCD00004880
n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowit
CHEBI:28939
CC(=O)NC(CS)C(=O)O

Spécifications

108°C to 111°C
616-91-1
C5H9NO3S
1724426
Soluble in water,ethanol,methanol,dimethyl sulfoxide,hot isopropyl alcohol,methyl acetate and ethyl acetate. Insoluble in chloroform and ether.
CC(=O)NC(CS)C(=O)O
100 g
12035
163.2
N-Acetyl-L-cysteine
≥98%
MFCD00004880
n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowit
PWKSKIMOESPYIA-BYPYZUCNSA-N
(2R)-2-acetamido-3-sulfanylpropanoic acid
163.191
CHEBI:28939
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

EINECSNumber : 210-498-3

RTECSNumber : HA1660000

TSCA : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions