missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-alanine, 96%

Catalog No. p-7045738
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1181103 1 g
AAL1181106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1181103 Supplier Thermo Scientific Chemicals Supplier No. L1181103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97-69-8 | C5H9NO3 | 131.131 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-69-8
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.131
MDL Number MFCD00063132
InChI Key KTHDTJVBEPMMGL-VKHMYHEASA-N
Synonym n-acetyl-l-alanine, ac-ala-oh, acetylalanine, acetyl-l-alanine, n-acetylalanine, 2-acetamidopropionic acid, l-n-acetylalanine, n-acetyl-s-alanine, l-alanine, n-acetyl, 2s-2-acetamidopropanoic acid
PubChem CID 88064
ChEBI CHEBI:40992
IUPAC Name (2S)-2-acetamidopropanoic acid
SMILES CC(C(=O)O)NC(=O)C

Specifications

Melting Point 122°C to 128°C
Beilstein 1722932
Quantity 1 g
Formula Weight 131.13
Optical Rotation −64° (c=2 in Water)
Percent Purity 96%
Chemical Name or Material N-Acetyl-L-alanine
EINECSNumber 202-602-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only