Learn More
N-Acetyl-β-D-mannosamine Hydrate, MP Biomedicals™
N-Acetyl-β-D-mannosamine Hydrate is an essential precursor of N-acetylneuraminic acid (NeuAc).
Supplier:  MP Biomedicals Inc 0210010905
 View more versions of this product
Catalog No. ICN10010905
Only null left
Description
Description
  • N-Acetyl-β-D-mannosamine hydrate is used as a substrate to identify, differentiate and characterize enzymes such as N-acetyl-glucosamine epimerase(s) and aldolase(s).
  • N-Acetyl-D-mannosamine is used for the synthesis of of N-acetylneuraminic acid.
Specifications
Specifications
White to Off-white
5 g
>98%
Crystalline
N-acetyl-β-D-Mannosamine
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.