missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-β-D-mannosamine Hydrate, MP Biomedicals™

Catalog No. p-4403621
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10010901 1 g
ICN10010905 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10010901 Supplier MP Biomedicals Inc Supplier No. 0210010901

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

N-Acetyl-β-D-mannosamine Hydrate is an essential precursor of N-acetylneuraminic acid (NeuAc).

  • N-Acetyl-β-D-mannosamine hydrate is used as a substrate to identify, differentiate and characterize enzymes such as N-acetyl-glucosamine epimerase(s) and aldolase(s).
  • N-Acetyl-D-mannosamine is used for the synthesis of of N-acetylneuraminic acid.

Specifications

Color White to Off-white
Form Crystalline
Quantity 1 g
Product Type N-acetyl-β-D-Mannosamine
Purity or Quality Grade >98%

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.