missing translation for 'onlineSavingsMsg'
Learn More

N-acetyl-D-galactosamine, MP Biomedicals™

Catalog No. ICN10006425
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10006425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10006425 Supplier MP Biomedicals Inc Supplier No. 0210006425

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

N-Acetyl-D-galactosamine(GalNAc), an aminosugar, is a component of many O-linked and N-linked glycan structures

Chemical Identifiers

CAS 14215-68-0
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.209
MDL Number MFCD00065372
InChI Key OVRNDRQMDRJTHS-JAJWTYFOSA-N
Synonym n-acetyl-d-galactosamine, galnac, 2-acetamido-2-deoxy-d-galactopyranose, d-galnac, 2-acetamido-2-deoxy-d-galactose, n-acetyl-d-chondrosamine, n-3r,4r,5r,6r-2,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-3-yl acetamide, n-acetylgalactosamine, acetylgalactosamine, n-acetyl-galactosamine
PubChem CID 440552
ChEBI CHEBI:28497
IUPAC Name N-[(2R,3R,4R,5R,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)NC1C(C(C(OC1O)CO)O)O

Specifications

Color White
Quantity 25 g
Formula Weight 221.21
Physical Form Powder
Chemical Name or Material N-acetyl-D-galactosamine