missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide

Catalog No. AAJ65878MB
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65878MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65878MB Supplier Thermo Scientific™ Supplier No. J65878MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 189950-66-1 | C26H34N6O13, C26H34N6O13 | 638.59 g/mol

N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide is an chromogenic substrate for Caspase 3 and CCP-32.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 189950-66-1
Molecular Formula C26H34N6O13
Synonym Ac-DEVD-pNA

Specifications

Quantity 25 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) A colorimetric substrate for caspase-3
Formula Weight 638.58g/mol
Product Type Signal Transduction Reagent
Form Powder

RUO – Research Use Only