missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine 98.0+%, TCI America™

Catalog No. F07525G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07525G 5 g
F07521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07525G Supplier TCI America Supplier No. F07525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 167015-11-4
Molecular Formula C37H31NO4S
Molecular Weight (g/mol) 585.718
MDL Number MFCD00151922
InChI Key KLBPUVPNPAJWHZ-UUWRZZSWSA-N
Synonym fmoc-d-cys trt-oh, n-fmoc-s-trityl-d-cysteine, fmoc-s-trityl-d-cysteine, n-alpha-9-fluorenylmethyloxycarbonyl-s-trityl-d-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl-d-cysteine, d-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tritylthio propanoic acid, ambotzfaa1035, pubchem19012
PubChem CID 7168037
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-tritylsulfanylpropanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 5 g
Formula Weight 585.72
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine
TSCA No
Recommended Storage Refrigerator