missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-pentafluoro-L-phenylalanine 97.0+%, TCI America™

Catalog No. F0900200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0900200MG 200 mg
F09001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0900200MG Supplier TCI America Supplier No. F0900200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 205526-32-5
Molecular Formula C24H16F5NO4
Molecular Weight (g/mol) 477.39
MDL Number MFCD00270612
InChI Key DLOGILOIJKBYKA-KRWDZBQOSA-N
Synonym fmoc-pentafluoro-l-phenylalanine, fmoc-phe f5-oh, fmoc-l-pentafluorophenylalanine, fmoc-l-pentafluorophe, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-perfluorophenyl propanoic acid, fmoc-3-pentafluorophenyl-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-pentafluoro-l-phenylalanine, n-alpha-9-fluorenylmethyloxycarbonyl-l-pentafluoro-phenylalanine, s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-pentafluorophenyl-propionic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-pentafluorophenyl propanoic acid
PubChem CID 7020337
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2,3,4,5,6-pentafluorophenyl)propanoic acid
SMILES OC(=O)[C@H](CC1=C(F)C(F)=C(F)C(F)=C1F)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White
Quantity 200 mg
Formula Weight 477.39
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-pentafluoro-L-phenylalanine
TSCA No
Recommended Storage Refrigerator