missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-valine 98.0+%, TCI America™

Catalog No. F02995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02995G 5 g
F029925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02995G Supplier TCI America Supplier No. F02995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68858-20-8
Molecular Formula C20H21NO4
Molecular Weight (g/mol) 339.39
MDL Number MFCD00037124
InChI Key UGNIYGNGCNXHTR-SFHVURJKSA-N
Synonym fmoc-l-valine, fmoc-val-oh, fmoc-l-val-oh, n-fmoc-l-valine, n-9h-fluoren-9-ylmethoxy carbonyl-l-valine, n-9-fluorenylmethoxycarbonyl-l-valine, fmoc-valine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylbutanoic acid, n-alpha-fmoc-l-valine, s-n-9h-fluoren-9-ylmethoxy carbonyl valine
PubChem CID 688217
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylbutanoic acid
SMILES CC(C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Melting Point 145°C
Color White
Quantity 5 g
Formula Weight 339.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-valine