missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine 95.0+%, TCI America™

Catalog No. F04565G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04565G 5 g
F04561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04565G Supplier TCI America Supplier No. F04565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92954-90-0
Molecular Formula C24H21NO5
Molecular Weight (g/mol) 403.43
MDL Number MFCD00134890
InChI Key SWZCTMTWRHEBIN-ANBDAQEENA-N
Synonym fmoc-tyr-oh, fmoc-l-tyrosine, fmoc-tyrosine, nalpha-fmoc-l-tyrosine, n-fmoc-l-tyrosine, fmoc-tyr, n-9h-fluoren-9-ylmethoxy carbonyl-l-tyrosine, n, a-fmoc-l-tyrosine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-hydroxyphenyl propanoic acid, tyrosine, n-9h-fluoren-9-ylmethoxy carbonyl
PubChem CID 6957986
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-hydroxyphenyl)propanoic acid
SMILES OC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White
Quantity 5 g
Formula Weight 403.43
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine