missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline 98.0+%, TCI America™
Supplier: TCI America F029825G
Specifications
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline | |
| 118°C | |
| C20H19NO4 | |
| 25 g | |
| ZPGDWQNBZYOZTI-UHFFFAOYNA-N | |
| 1-{[(9H-fluoren-9-yl)methoxy]carbonyl}pyrrolidine-2-carboxylic acid | |
| 688135 | |
| ≥98.0% (HPLC,T) |
| 71989-31-6 | |
| White | |
| MFCD00037122 | |
| fmoc-pro-oh, fmoc-l-proline, fmoc-l-pro-oh, n-fmoc-l-proline, n-9-fluorenylmethoxycarbonyl-l-proline, s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carboxylic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-proline, fmoc-pro, 2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, 9-fluorenylmethoxycarbonyl-l-proline | |
| OC(=O)C1CCCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 337.38 | |
| 337.38 | |
| Crystalline Powder |
Chemical Identifiers
| 71989-31-6 | |
| 337.38 | |
| ZPGDWQNBZYOZTI-UHFFFAOYNA-N | |
| 688135 | |
| OC(=O)C1CCCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| C20H19NO4 | |
| MFCD00037122 | |
| fmoc-pro-oh, fmoc-l-proline, fmoc-l-pro-oh, n-fmoc-l-proline, n-9-fluorenylmethoxycarbonyl-l-proline, s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carboxylic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-proline, fmoc-pro, 2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, 9-fluorenylmethoxycarbonyl-l-proline | |
| 1-{[(9H-fluoren-9-yl)methoxy]carbonyl}pyrrolidine-2-carboxylic acid |
Safety and Handling
TSCA : No