missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline 98.0+%, TCI America™

Catalog No. p-7129598
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02985G 5 g
F029825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02985G Supplier TCI America Supplier No. F02985G
Only null left

Chemical Identifiers

CAS 71989-31-6
Molecular Formula C20H19NO4
Molecular Weight (g/mol) 337.38
MDL Number MFCD00037122
InChI Key ZPGDWQNBZYOZTI-UHFFFAOYNA-N
Synonym fmoc-pro-oh, fmoc-l-proline, fmoc-l-pro-oh, n-fmoc-l-proline, n-9-fluorenylmethoxycarbonyl-l-proline, s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carboxylic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-proline, fmoc-pro, 2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, 9-fluorenylmethoxycarbonyl-l-proline
PubChem CID 688135
IUPAC Name 1-{[(9H-fluoren-9-yl)methoxy]carbonyl}pyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 118°C
Color White
Quantity 5 g
Formula Weight 337.38
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline