accessibility menu, dialog, popup

UserName

HOONOO

CAS RN 71989-31-6

IUPAC Name: 1-{[(9H-fluoren-9-yl)methoxy]carbonyl}pyrrolidine-2-carboxylic acid
Synonyms: 1-[(9H-fluoren-9-ylmethoxy)carbonyl]pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 337.38
Molecular Formula: C20H19NO4
InChi Key: ZPGDWQNBZYOZTI-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)