missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine 98.0+%, TCI America™

Catalog No. p-7129823
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06691G 1 g
F06695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06691G Supplier TCI America Supplier No. F06691G
Only null left

Chemical Identifiers

CAS 102410-65-1
Molecular Formula C23H19NO4
Molecular Weight (g/mol) 373.408
MDL Number MFCD00155632
InChI Key PCJHOCNJLMFYCV-NRFANRHFSA-N
Synonym fmoc-phg-oh, fmoc-l-phenylglycine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-phenylacetic acid, n-fmoc-l-2-phenylglycine, fmoc-s-phenylglycine, n-9h-fluoren-9-ylmethoxy carbonyl-l-2-phenylglycine, n-alpha-9-fluorenylmethyloxycarbonyl-l-phenylglycine, s-9h-fluoren-9-ylmethoxycarbonylamino-phenyl-acetic acid, s-9h-fluoren-9-ylmethoxy carbonyl amino phenyl acetic acid, 2s-9h-fluoren-9-yl-methoxy carbonyl amino phenyl acetic acid
PubChem CID 7269367
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 1 g
Formula Weight 373.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine