missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-serine 95.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 112°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 327.34 |
| Percent Purity | ≥95.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-serine |
Sécurité et manipulation
| TSCA | No |