missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine 98.0+%, TCI America™
Supplier: TCI America F1013200MG
Specifications
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine | |
| 139°C | |
| C22H19NO5 | |
| 200 mg | |
| AJXDCHXGNUFBRC-FQEVSTJZSA-N | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(furan-2-yl)propanoic acid | |
| 2734451 | |
| ≥98.0% (T) |
| 159611-02-6 | |
| White-Yellow | |
| MFCD01311747 | |
| fmoc-l-2-furylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-beta-2-furyl-ala-oh, fmoc-3-2-furyl-l-alanine, fmoc-l-3-2-furyl-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-3-2-furyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-furan-2-yl propanoic acid, rarechem bk pt 0223, s-n-fmoc-furylalanine, fmoc-3-ala 2-furyl-oh | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CO4)C(=O)O | |
| 377.396 | |
| 377.40 | |
| Crystalline Powder |
Chemical Identifiers
| 159611-02-6 | |
| 377.396 | |
| AJXDCHXGNUFBRC-FQEVSTJZSA-N | |
| 2734451 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CO4)C(=O)O |
| C22H19NO5 | |
| MFCD01311747 | |
| fmoc-l-2-furylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-beta-2-furyl-ala-oh, fmoc-3-2-furyl-l-alanine, fmoc-l-3-2-furyl-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-3-2-furyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-furan-2-yl propanoic acid, rarechem bk pt 0223, s-n-fmoc-furylalanine, fmoc-3-ala 2-furyl-oh | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(furan-2-yl)propanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator