missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine 98.0+%, TCI America™

Catalog No. p-7130027
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1013200MG 200 mg
F10131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1013200MG Supplier TCI America Supplier No. F1013200MG

Chemical Identifiers

CAS 159611-02-6
Molecular Formula C22H19NO5
Molecular Weight (g/mol) 377.396
MDL Number MFCD01311747
InChI Key AJXDCHXGNUFBRC-FQEVSTJZSA-N
Synonym fmoc-l-2-furylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-beta-2-furyl-ala-oh, fmoc-3-2-furyl-l-alanine, fmoc-l-3-2-furyl-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-3-2-furyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-furan-2-yl propanoic acid, rarechem bk pt 0223, s-n-fmoc-furylalanine, fmoc-3-ala 2-furyl-oh
PubChem CID 2734451
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(furan-2-yl)propanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CO4)C(=O)O

Specifications

Melting Point 139°C
Color White-Yellow
Quantity 200 mg
Formula Weight 377.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine
TSCA No
Recommended Storage Refrigerator