missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Chlorophenyl)phthalimide 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 198°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 257.67 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-(4-Chlorophenyl)phthalimide |
Sécurité et manipulation
| RTECSNumber | TI4267000 |
| TSCA | No |