missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Chlorophenyl)formamide 98.0+%, TCI America™

Catalog No. C19455G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C19455G Supplier TCI America Supplier No. C19455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2617-79-0
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.581
MDL Number MFCD00021020
InChI Key LMLFHXMNNHGRRO-UHFFFAOYSA-N
Synonym 1-Chloro-4-formamidobenzene, 4′C-Chloroformanilide
PubChem CID 17475
IUPAC Name N-(4-chlorophenyl)formamide
SMILES C1=CC(=CC=C1NC=O)Cl

Specifications

Melting Point 104°C
Color White
Quantity 5 g
Formula Weight 155.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Chlorophenyl)formamide
RTECSNumber LQ4666000
TSCA No