missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide 98.0+%, TCI America™

Catalog No. C33155G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33155G 5 g
C33151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33155G Supplier TCI America Supplier No. C33155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 848133-76-6
Molecular Formula C14H12ClN3O2
Molecular Weight (g/mol) 289.72
MDL Number MFCD09263704
InChI Key XDXGFTCQRAQEEG-UHFFFAOYSA-N
PubChem CID 11358234
IUPAC Name N-(4-chloro-3-cyano-7-ethoxyquinolin-6-yl)acetamide
SMILES CCOC1=C(NC(C)=O)C=C2C(Cl)=C(C=NC2=C1)C#N

Specifications

Melting Point 250°C
Quantity 5 g
Formula Weight 289.72
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide