missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromophenyl)phthalimide 98.0+%, TCI America™

Catalog No. B27075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27075G 5 g
B270725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27075G Supplier TCI America Supplier No. B27075G
Only null left

Chemical Identifiers

CAS 40101-31-3
Molecular Formula C14H8BrNO2
Molecular Weight (g/mol) 302.13
MDL Number MFCD00047301
InChI Key DNQCPYWSXMATIN-UHFFFAOYSA-N
PubChem CID 101305
IUPAC Name 2-(4-bromophenyl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES BrC1=CC=C(C=C1)N1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 206°C
Color White
Quantity 5 g
Formula Weight 302.13
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Bromophenyl)phthalimide
RTECSNumber TI4025000
TSCA No