missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromophenyl)-3-(trifluoromethyl)benzamide, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H5701503

 View more versions of this product

Catalog No. AAH5701503


Only null left
Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

N-(4-Bromophenyl)-3-(trifluoromethyl)benzamide
C14H9BrF3NO
1 g
n-4-bromophenyl-3-trifluoromethyl benzamide, benzamide, n-4-bromophenyl-3-trifluoromethyl, n-4-bromo-phenyl-3-trifluoromethyl-benzamide, n-4-bromophenyl 3-trifluoromethyl phenyl carboxamide
FC(F)(F)C1=CC=CC(=C1)C(=O)NC1=CC=C(Br)C=C1
344.13
344.13
562080-79-9
MFCD03937564
14213316
JDUHFWWSDVISOZ-UHFFFAOYSA-N
N-(4-bromophenyl)-3-(trifluoromethyl)benzamide
531810
97%

Chemical Identifiers

562080-79-9
344.13
JDUHFWWSDVISOZ-UHFFFAOYSA-N
531810
FC(F)(F)C1=CC=CC(=C1)C(=O)NC1=CC=C(Br)C=C1
C14H9BrF3NO
MFCD03937564
n-4-bromophenyl-3-trifluoromethyl benzamide, benzamide, n-4-bromophenyl-3-trifluoromethyl, n-4-bromo-phenyl-3-trifluoromethyl-benzamide, n-4-bromophenyl 3-trifluoromethyl phenyl carboxamide
N-(4-bromophenyl)-3-(trifluoromethyl)benzamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions