CAS RN 562080-79-9
IUPAC Name:
N-(4-bromophenyl)-3-(trifluoromethyl)benzamide
Synonyms:
N-(4-bromophenyl)-3-(trifluoromethyl)benzamide
Molecular Weight (g/mol):
344.13
Molecular Formula:
C14H9BrF3NO
InChi Key:
JDUHFWWSDVISOZ-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=CC(=C1)C(=O)NC1=CC=C(Br)C=C1
1
–
1
of
1
results
1
N-(4-Bromophenyl)-3-(trifluoromethyl)benzamide, 97%, Thermo Scientific™
CAS: 562080-79-9 Molecular Formula: C14H9BrF3NO Molecular Weight (g/mol): 344.13 MDL Number: MFCD03937564 InChI Key: JDUHFWWSDVISOZ-UHFFFAOYSA-N Synonym: n-4-bromophenyl-3-trifluoromethyl benzamide,benzamide, n-4-bromophenyl-3-trifluoromethyl,n-4-bromo-phenyl-3-trifluoromethyl-benzamide,n-4-bromophenyl 3-trifluoromethyl phenyl carboxamide PubChem CID: 531810 IUPAC Name: N-(4-bromophenyl)-3-(trifluoromethyl)benzamide SMILES: FC(F)(F)C1=CC=CC(=C1)C(=O)NC1=CC=C(Br)C=C1