missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromophenyl)-3-(trifluoromethyl)benzamide, 97%, Thermo Scientific™

Catalog No. p-7038116
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5701503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5701503 Supplier Thermo Scientific Chemicals Supplier No. H5701503
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 562080-79-9
Molecular Formula C14H9BrF3NO
Molecular Weight (g/mol) 344.13
MDL Number MFCD03937564
InChI Key JDUHFWWSDVISOZ-UHFFFAOYSA-N
Synonym n-4-bromophenyl-3-trifluoromethyl benzamide, benzamide, n-4-bromophenyl-3-trifluoromethyl, n-4-bromo-phenyl-3-trifluoromethyl-benzamide, n-4-bromophenyl 3-trifluoromethyl phenyl carboxamide
PubChem CID 531810
IUPAC Name N-(4-bromophenyl)-3-(trifluoromethyl)benzamide
SMILES FC(F)(F)C1=CC=CC(=C1)C(=O)NC1=CC=C(Br)C=C1

Specifications

Quantity 1 g
Beilstein 14213316
Formula Weight 344.13
Percent Purity 97%
Chemical Name or Material N-(4-Bromophenyl)-3-(trifluoromethyl)benzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures