missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromobutyl)phthalimide 95.0+%, TCI America™

Catalog No. B13945G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13945G 5 g
B139425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13945G Supplier TCI America Supplier No. B13945G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5394-18-3
Molecular Formula C12H12BrNO2
Molecular Weight (g/mol) 282.14
MDL Number MFCD00005905
InChI Key UXFWTIGUWHJKDD-UHFFFAOYSA-N
Synonym n-4-bromobutyl phthalimide, 2-4-bromobutyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-bromobutyl, 2-4-bromobutyl-1h-isoindole-1,3 2h-dione, n-4-bromobutyl-phthalimide, 2-4-bromobutyl isoindole-1,3-dione, 2-4-bromobutyl-2,3-dihydro-1h-isoindole-1,3-dione, n-4-bromobutyl, n-bromobutylphthalimide
PubChem CID 93575
IUPAC Name 2-(4-bromobutyl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES BrCCCCN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 5 g
Formula Weight 282.14
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Bromobutyl)phthalimide