missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Aminobutyl)-N-ethylisoluminol 97.0+%, TCI America™

Catalog No. A53041G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A53041G 1 g
A5304100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A53041G Supplier TCI America Supplier No. A53041G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66612-29-1
Molecular Formula C14H20N4O2
Molecular Weight (g/mol) 276.34
MDL Number MFCD00010559
InChI Key LEOJISUPFSWNMA-UHFFFAOYSA-N
Synonym n-4-aminobutyl-n-ethylisoluminol, abei, 6-4-aminobutyl ethyl amino-2,3-dihydrophthalazine-1,4-dione, 6-n-4-aminobutyl-n-ethyl amino-2,3-dihydrophthalazine-1,4-dione, 1,4-phthalazinedione, 6-4-aminobutyl ethylamino-2,3-dihydro, 6-n-4-aminobutyl-n-ethylamino-2,3-dihydro-1,4-phthalazinedione, n-4-aminobutyl-n-ethylisolumiol
PubChem CID 196441
IUPAC Name 6-[(4-aminobutyl)(ethyl)amino]-1,2,3,4-tetrahydrophthalazine-1,4-dione
SMILES CCN(CCCCN)C1=CC=C2C(=O)NNC(=O)C2=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 276.34
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Aminobutyl)-N-ethylisoluminol [Chemiluminescence Reagent]