missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Aminobutyl)-5-chloronaphthalene-2-sulfonamide Hydrochloride 98.0+%, TCI America™

Catalog No. p-7117893
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2416500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2416500MG Supplier TCI America Supplier No. A2416500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88519-57-7
Molecular Formula C14H18Cl2N2O2S
Molecular Weight (g/mol) 349.27
MDL Number MFCD00058030
InChI Key QKAALLVQBOLELJ-UHFFFAOYSA-N
Synonym w-13 hydrochloride, n-4-aminobutyl-5-chloronaphthalene-2-sulfonamide hydrochloride, w-13, hydrochloride, n-4-aminobutyl-5-chloro-2-naphthalenesulfonamide hydrochloride, 2-naphthalenesulfonamide,n-4-aminobutyl-5-chloro-, hydrochloride 1:1, n-4-aminobutyl-5-chloro-2-naphthalenesulfonamide, hcl, acmc-209qtx, n-4-aminobutyl-5-chloro-2-naphthalenesulfonamide hcl, n-4-aminobutyl-5-chloronaphthalene-2-sulfonamidehydrochloride, n-4-aminobutyl-5-chloro-2-naphthalene sulfonamide hydro chloride
PubChem CID 16760706
ChEBI CHEBI:75367
IUPAC Name N-(4-aminobutyl)-5-chloronaphthalene-2-sulfonamide;hydrochloride
SMILES C1=CC2=C(C=CC(=C2)S(=O)(=O)NCCCCN)C(=C1)Cl.Cl

Specifications

Melting Point 237°C
Quantity 500 mg
Formula Weight 349.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Aminobutyl)-5-chloronaphthalene-2-sulfonamide Hydrochloride
TSCA No
Recommended Storage Refrigerator